Order Entry
Puerto Rico
ContactUsLinkComponent
Multiple Species Recombinant DHA (from E. coli), Ovalbumin
Multiple Species Recombinant DHA (from E. coli), Ovalbumin
Catalog # MSPP-CPO623GE1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Multiple Species Recombinant DHA (from E. coli), Ovalbumin
Catalog # MSPP-CPO623GE1
Supplier:  Cloud-Clone
Supplier Number:  CPO623GE11
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Ovalbumin
  • Protein Function:
    Bio-Markers & CD Antigens
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Multiple
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Docosahexaenoic Acid
  • Protein/Peptide Name:
    DHA
  • Purity:
    90 - 100%
  • Molecular Weight:
    328.5 Da
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-CPO623GE1

Specifications

About this item

This is a DHA conjugated BSA, General species is sequencing from - with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Docosahexaenoic acid (DHA) is an omega-3 fatty acid that is a primary structural component of the human brain, cerebral cortex, skin, sperm, testicles and retina. It can be synthesized from alpha-linolenic acid or obtained directly from maternal milk or fish oil. DHA's structure is a carboxylic acid (~oic acid) with a 22-carbon chain (docosa- is Greek for 22) and six (Greek 'hexa') cis double bonds (-en~); the first double bond is located at the third carbon from the omega end. Cold-water oceanic fish oils are rich in DHA. Most of the DHA in fish and multi-cellular organisms with access to cold-water oceanic foods originates from photosynthetic and heterotrophic microalgae, and becomes increasingly concentrated in organisms the further they are up the food chain. DHA is also commercially manufactured from microalgae; Crypthecodinium cohnii and another of the genus Schizochytrium. [4] DHA manufactured using microalgae is vegetarian.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.