Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant SPINK5 (prokaryotic) (from E.coli)
  MSPP-RPH145MU1
 :  
undefined
Mouse Recombinant SPINK5 (prokaryotic) (from E.coli)
  MSPP-RPH145MU1
 :  RPH145MU01
 :  

 

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    72432
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Serine Peptidase Inhibitor Kazal Type 5
  • UniProtKB:
    Q148R4
  • Protein/Peptide Name:
    SPINK5 (prokaryotic)
  • Purity:
    90 - 100%
  • Molecular Weight:
    36 kDa
  • Sequence:
    Cys734~Ala995
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPH145MU1

 

 

This is a SPINK5 recombinant protein (prokaryotic), Mouse is sequencing from Cys734~Ala995 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

SPINK5 encodes a multidomain serine protease inhibitor that contains 15 potential inhibitory domains. The inhibitor may play a role in skin and hair morphogenesis and anti-inflammatory and/or antimicrobial protection of mucous epithelia. Mutations may result in Netherton syndrome, a disorder characterized by ichthyosis, defective cornification, and atopy. The entire precursor protein, as deduced from the nucleotide sequence of the cloned cDNA, includes 15 potential inhibitory domains, 13 of which exhibit a Kazal-type-derived 4-cysteine-residue pattern that represents a novel protein module of serine protease inhibitor. Northern blot analysis and RT-PCR detected expression of LEKTI in thymus, vaginal epithelium, Bartholin gland, oral mucosa, tonsil, and parathyroid gland.

 : ISO 9001 and ISO 13485 certified, three level QC system.
 : For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.