Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant TLR9 (prokaryotic) (from E.coli)
Human Recombinant TLR9 (prokaryotic) (from E.coli)
Catalog # MSPP-RPA709HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant TLR9 (prokaryotic) (from E.coli)
Catalog # MSPP-RPA709HU1
Supplier:  Cloud-Clone
Supplier Number:  RPA709HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    54106
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Toll Like Receptor 9
  • UniProtKB:
    Q9NR96
  • Protein/Peptide Name:
    TLR9 (prokaryotic)
  • Purity:
    97 - 100%
  • Molecular Weight:
    41 kDa
  • Sequence:
    Thr28~Ser362
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA709HU1

Specifications

About this item

This is a TLR9 recombinant protein (prokaryotic), Human is sequencing from Thr28~Ser362 with 97 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Toll-like receptors (TLRs) recognize microbial products and initiate a complex immune response designed to eliminate invading pathogens. TLR9 is specialized to recognize microbial nucleic acids, and 1 factor enabling TLR9 to discriminate between self and foreign DNA is the presence of unmethylated CpG dinucleotides in bacterial DNA (Hasan et al. 2007).other TLR proteins, TLR9 contains extracellular leucine-rich repeats (LRRs) and a cytoplasmic toll/interleukin-1R (TIR) domain. It also has a signal peptide (residues 1 to 25) and a transmembrane domain (residues 819 to 836). Northern blot analysis of mouse tissues revealed highest Tlr9 expression in spleen.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.