Order Entry
Puerto Rico
ContactUsLinkComponent
Multiple Species Recombinant H6 (from Synthetic), Ovalbumin
Multiple Species Recombinant H6 (from Synthetic), Ovalbumin
Catalog # MSPP-CPX656G21
Supplier:  Cloud-Clone
CAS Number:  
Multiple Species Recombinant H6 (from Synthetic), Ovalbumin
Catalog # MSPP-CPX656G21
Supplier:  Cloud-Clone
Supplier Number:  CPX656GE21
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Ovalbumin
  • Protein Function:
    Bio-Markers & CD Antigens
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Synthetic
  • Species:
    Multiple
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Hexa Histidine
  • Protein/Peptide Name:
    H6
  • Purity:
    90 - 100%
  • Sequence:
    HHHHHH
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-CPX656G21

Specifications

About this item

This is a H6 conjugated OVA, General species is sequencing from HHHHHH with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

The hexahistidine (His(6))/nickel(II)-nitrilotriacetic acid (Ni(2+)-NTA) system is widely used for affinity purification of recombinant proteins. The NTA group has many other applications, including the attachment of chromophores, fluorophores, or nanogold to His(6) proteins. Its high specificity makes (Ni(2+)-NTA)(2)-Cy3 ideal for detecting His(6) proteins in complex mixtures of other proteins, allowing (Ni(2+)-NTA)(2)-Cy3 to be used as a probe in crude cell extracts and as a His(6)-specific gel stain. (Ni(2+)-NTA)(2)-Cy3 binding is reversible in 10 mM ethylenediaminetetraacetic acid (EDTA) or 500 mM imidazole, but in their absence it exchanges slowly. Labeling with (Ni(2+)-NTA)(2)-Cy3 allows characterization of hydrodynamic properties by fluorescence anisotropy or analytical ultracentrifugation under conditions that prevent direct detection of protein.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.