- Pack type:Vial
- Conjugation:BSA (Bovine Serum Albumin)
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:Synthetic
- Species:Multiple
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:−20 °C
- Endotoxin Content:<1.0 EU per 1 µg (determined by the LAL method)
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Arachidonic Acid
- Protein/Peptide Name:AA
- Purity:90 - 100%
- Molecular Weight:304.5 Da
- Sequence:C20H32O2
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-CPB098GE1
This is a AA conjugated BSA, General species is sequencing from C20H32O2 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Arachidonic acid (AA, sometimes ARA) is an omega-6 fatty acid 20:4(ω-6). It is the counterpart to the saturated arachidic acid found in peanut oil, In chemical structure, arachidonic acid is a carboxylic acid with a 20-carbon chain and four cis double bonds; the first double bond is located at the sixth carbon from the omega end. Some chemistry sources define 'arachidonic acid' to designate any of the eicosatetraenoic acids. However, almost all writings in biology, medicine and nutrition limit the term to all-cis 5,8,11,14-eicosatetraenoic acid. Arachidonic acid is a polyunsaturated fatty acid that is present in the phospholipids (especially phosphatidylethanolamine, phosphatidylcholine and phosphatidylinositides) of membranes of the body's cells, and is abundant in the brain and muscles.