Order Entry
Puerto Rico
ContactUsLinkComponent
Multiple Species Recombinant E2 (from Synthetic), BSA (Bovine Serum Albumin)
Multiple Species Recombinant E2 (from Synthetic), BSA (Bovine Serum Albumin)
Catalog # MSPP-CPA461GE1
CAS Number:  
undefined
Multiple Species Recombinant E2 (from Synthetic), BSA (Bovine Serum Albumin)
Catalog # MSPP-CPA461GE1
Supplier Number:  CPA461GE11
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    BSA (Bovine Serum Albumin)
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Synthetic
  • Species:
    Multiple
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Estradiol
  • Protein/Peptide Name:
    E2
  • Purity:
    90 - 100%
  • Molecular Weight:
    272.4Da
  • Sequence:
    C18H24O2
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-CPA461GE1

Specifications

About this item

This is a E2 conjugated BSA, General species is sequencing from C18H24O2 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Estradiol is an estrogenic hormone, C18H24O2, produced by the ovaries and used in treating estrogen deficiency. Estradiol is essential for maintaining normal female functions and is normally produced by the ovaries. After menopause, the ovaries decrease their production of this hormone. Estradiol is a sex hormone. Estradiol is the predominant sex hormone present in females; however, it is present in males, albeit at lower levels, as well. It represents the major estrogen in humans. Estradiol has not only a critical impact on reproductive and sexual functioning, but also affects other organs including the bones. Estradiol is produced not only in the gonads: In both sexes, precursor hormones, to be specific testosterone, are converted by aromatization to estradiol. In particular, fat cells are active to convert precursors to estradiol, and will continue to do so even after menopause.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.