- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Chicken
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:−20 °C
- Endotoxin Content:<1.0 EU per 1 µg (determined by the LAL method)
- Gene ID:396413
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Fibroblast Growth Factor 2, Basic
- UniProtKB:P48800
- Protein/Peptide Name:FGF2 (prokaryotic)
- Purity:95 - 100%
- Molecular Weight:17/20 kDa
- Sequence:Phe29~Gly143
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPA551GA1
This is a FGF2 recombinant protein (prokaryotic), Chicken is sequencing from Phe29~Gly143 with 95 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
FGF basic is one of at least 22 mitogenic proteins of the FGF family, which show 35 to 60% amino acid conservation. Unlike other FGFs, FGF acidic and basic lack signal peptides and are secreted by an alternate pathway. Storage pools within the cell or on cell surface heparan sulfate proteoglycans (HSPG) are likely. The predicted 18 kDa FGF basic isoform can be located in both the cytoplasm and the nucleus and is presumed to be the form secreted. Transcription from alternate start sites produces 21 to 23 kDa forms found only in the nucleus. High and low molecular weight human FGF basic targets the expression of different genes when expressed in NIH-3T3 cells. The 18 kDa rat sequence has 98% aa identity with mouse, and 96 to 97% identity with human, bovine and sheep FGF basic.