- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:−20 °C
- Endotoxin Content:<1.0 EU per 1 µg (determined by the LAL method)
- Gene ID:3458
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Interferon Gamma
- UniProtKB:P01579
- Protein/Peptide Name:IFNg (prokaryotic)
- Purity:95 - 100%
- Molecular Weight:16 kDa
- Sequence:Gln24~Gln166
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPA049HU1
This is a IFNg recombinant protein (prokaryotic), Human is sequencing from Gln24~Gln166 with 95 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Interferon gamma (IFN-gama ) is a multifunctional protein first observed as an antiviral activity in cultures of Sindbis virus-infected leukocytes stimulated by PHA. Produced by T lymphocytes and natural killer (NK) cells, IFN-γ is now known to be both an inhibitor of viral replication and a regulator of numerous immunological functions. IFN-γ influences the class of antibody produced by B cells, upregulates class I and II MHC complex antigens and increases. The efficiency of macrophage-mediated killing of intracellular parasites. Most of the activities attributed to IFN-γ are believed to be mediated by IFN-γ-induced proteins. The appearance of such proteins is a consequence of IFN-γ binding to a specific receptor that is distinct from the receptor for IFN-αandβ.