Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant NT-ProBNP (prokaryotic) (from E.coli)
Human Recombinant NT-ProBNP (prokaryotic) (from E.coli)
Catalog # MSPP-RPA485HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant NT-ProBNP (prokaryotic) (from E.coli)
Catalog # MSPP-RPA485HU1
Supplier:  Cloud-Clone
Supplier Number:  RPA485HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    4879
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    N-Terminal Pro-Brain Natriuretic Peptide
  • UniProtKB:
    P16860
  • Protein/Peptide Name:
    NT-ProBNP (prokaryotic)
  • Purity:
    97 - 100%
  • Molecular Weight:
    12 kDa
  • Sequence:
    His27~Arg102
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA485HU1

Specifications

About this item

This is a NT-ProBNP recombinant protein (prokaryotic), Human is sequencing from His27~Arg102 with 97 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Brain-type Natriuretic Peptide (BNP) is a nonglycosylated peptide that is produced predominantly by ventricular myocytes and belongs to the natriuretic peptide family. Plasma NT-proBNP is a marker for congestive heart failure, while mature BNP (aa 103 - 134) promotes vasodilation and fluid and sodium excretion. BNP is co-secreted along with a 76 amino acid N-terminal fragment (NT-proBNP) which is biologically inactive. BNP binds to and activates the atrial natriuretic factor receptors NPRA, and to a lesser extent NPRB, in a fashion similar to atrial natriuretic peptide (ANP) but with 10-fold lower affinity. The biological half-life of BNP, however, is twice as long as that of ANP, and that of NT-proBNP is even longer, making these peptides better targets than ANP for diagnostic blood testing.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.