Order Entry
Puerto Rico
ContactUsLinkComponent
Multiple Species Recombinant GFP (eukaryotic) (from Yeast)
Multiple Species Recombinant GFP (eukaryotic) (from Yeast)
Catalog # MSPP-EPD25GE51
CAS Number:  
undefined
Multiple Species Recombinant GFP (eukaryotic) (from Yeast)
Catalog # MSPP-EPD25GE51
Supplier Number:  EPD025GE51
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Bio-Markers & CD Antigens
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Yeast
  • Species:
    Multiple
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Reconstitution Instructions:
    Do not vortex
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Green Fluorescent Protein
  • UniProtKB:
    P42212
  • Protein/Peptide Name:
    GFP (eukaryotic)
  • Purity:
    95 - 100%
  • Molecular Weight:
    28 kDa
  • Sequence:
    Met1~Lys238
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Liquid with PBS, pH 7.4, containing 20% glycerine
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-EPD25GE51

Specifications

About this item

This is a GFP recombinant protein (eukaryotic,Yeast), General species is sequencing from Met1~Lys238 with 95 to 100% purity. Liquid with PBS, pH 7.4, containing 20 Glycerine with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

The green fluorescent protein (GFP) is a protein composed of 238 amino acid residues (26.9 kDa) that exhibits bright green fluorescence when exposed to light in the blue to ultraviolet range. Although many other marine organisms have similar green fluorescent proteins, GFP traditionally refers to the protein first isolated from the jellyfish Aequorea victoria. The GFP from A. victoria has a major excitation peak at a wavelength of 395 nm and a minor one at 475 nm. Its emission peak is at 509 nm, which is in the lower green portion of the visible spectrum. The GFP from the sea pansy has a single major excitation peak at 498 nm. In cell and molecular biology, the GFP gene is frequently used as a reporter of expression. In modified forms it has been used to make biosensors, and many animals have been created that express GFP as a proof-of-concept that a gene can be expressed throughout a given organism.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.