Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant GFAP (prokaryotic) (from E.coli)
Human Recombinant GFAP (prokaryotic) (from E.coli)
Catalog # MSPP-RPA068HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant GFAP (prokaryotic) (from E.coli)
Catalog # MSPP-RPA068HU1
Supplier:  Cloud-Clone
Supplier Number:  RPA068HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    2670
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Glial Fibrillary Acidic Protein
  • UniProtKB:
    P14136
  • Protein/Peptide Name:
    GFAP (prokaryotic)
  • Purity:
    90 - 100%
  • Molecular Weight:
    55 kDa
  • Sequence:
    Met1~Met432
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA068HU1

Specifications

About this item

This is a GFAP recombinant protein (prokaryotic), Human is sequencing from Met1~Met432 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

GFAP) is an intermediate filament (IF) protein that is found in glial cells such as astrocytes. It is closely related to its non-epithelial family members, vimentin, desmin, and peripherin, which are all involved in the structure and function of the cell’s cytoskeleton. GFAP helps to maintain astrocyte mechanical strength, as well as the shape of cells. GFAP is expressed in the central nervous system in astrocyte cells. It is involved in many cellular functioning processes, such as cell structure and movement, cell communication, and the functioning of the blood brain barrier. The predicted amino acid sequence indicated that GFAP shares structural similarities particularly in the central rod domain and to a lesser degree in the carboxyl-terminal domain with other IF proteins found in nonepithelial cell types.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.