- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:−20 °C
- Endotoxin Content:<1.0 EU per 1 µg (determined by the LAL method)
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Thrombospondin 1
- UniProtKB:P35441
- Protein/Peptide Name:THBS1 (prokaryotic)
- Purity:90 - 100%
- Molecular Weight:25 kDa
- Sequence:Ile61~Ile259
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPA611MU1
This is a THBS1 recombinant protein (prokaryotic), Mouse is sequencing from Ile61~Ile259 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Thrombospondin-1 (TSP-1) is a 150 to 180 kDa charter member of the thrombospondin family of extracellular matrix proteins. 1 - 4 Human TSP-1 is synthesized as an 1170 amino acid (aa) precursor that contains an 18 aa signal sequence and 1152 aa mature molecule. The mature molecule has been described as containing three distinct regions that create the shape of a dumbbell. There is an intital, 140 aa N-terminal laminin G-like globular region that binds heparin (aa 19 - 258). This is followed by an extended, central collagen-binding region that contains one type C von Willebrand factor domain, plus three TSP type I and three TSP type II (or EGF-like) domains (aa 259 - 712). The type I repeats have multiple functions. They bind to type IV collagen, laminin, fibronectin and CD36.