- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:−20 °C
- Endotoxin Content:<1.0 EU per 1 µg (determined by the LAL method)
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Immunoglobulin E
- UniProtKB:P06336
- Protein/Peptide Name:IgE (prokaryotic)
- Purity:90 - 100%
- Molecular Weight:51 kDa
- Sequence:Ser1~Ser421
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPA545MU1
This is a IgE recombinant protein (prokaryotic), Mouse is sequencing from Ser1~Ser421 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Immunoglobulin E (IgE) is a class of antibody produced in the lungs, skin, and mucous membranes that has only been found in mammals. It plays an important role in allergy, and is especially associated with type 1 hypersensitivity. IgE has also been implicated in immune system responses to most parasitic worms like Schistosoma mansoni, Trichinella spiralis, and Fasciola hepatica, and may be important during immune defense against certain protozoan parasites such as Plasmodium falciparum. The antigen-specific IgE interacts with mast cells and eosinophils, triggers the release of histamine, leukotrienes and other substances that lead to the itching, sneezing and congestion of allergies - and the life threatening respiratory distress of asthma and anaphylactic shock.