Specifications
- Antibody Type:Primary
- Antigen Name:BCMA CAR, scFv region of a BCMA-specific mouse mAb clone C11D5.3
- Antigen Symbol:C11D5.3 scFv
- Clonality:Monoclonal
- Clone:RA3
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Size:100 tests
- Storage Buffer:Aqueous buffered solution containing protein stabilizer and ≤0.05% ProClin 300
- Storage Temperature:At 2...8 °C, in dark place
- Concentration:1 μl per test, recommended
- Shipping Temperature:20
- Immunogen:scFv region of a BCMA-specific mouse mAb clone C11D5.3
- Purification:Protein A affinity purification
- Cat. No.:MSPP-200706
- Packaging:Vial
Specifications
About this item
1A3 specifically binds to the scFv region of a B-cell maturation antigen (BCMA) specific mouse monoclonal antibody (mAb, clone C11D5.3). BCMA is a protein that has been reported to be selectively expressed by B-lineage cells including multiple myeloma cells1 and restrictively expressed in both normal and malignant plasma cells at high levels2. The scFv region of C11D5.3 has been used to develop BCMA-specific chimeric antigen receptor (CAR) T cells utilized in clinical trials.
- High sensitivity
- Low non-specificity
- Idiotypic
- Diagnostic material for CAR-T CDx
Reliable CAR-T CDx, been in helping commercialized CAR-T therapeutics. Immediate shipping if in stock, lead time might be up to 30 days if out of stock pending for reproduction.
Type: Primary
Antigen: C11D5.3 scFv
Clonality: Monoclonal
Clone: RA3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse