KERLIX™ Sponges, Covidien
Supplier: Cardinal Health
Certificates
About this item
Prewashed, fluff-dried woven gauze has a crinkle-weave pattern for loft and bulk to cushion and protect wound areas.
- Made from high-grade 100% cotton
Absorbency and texture are ideal for prepping, cleansing, packing, and debriding wounds.
Certifications: Exceed USP Type VII gauze requirements.
Ordering information: Nonsterile sponges are bulk packaged. Sterile sponges are packaged 10 to a plastic tray.