
1-(2-Hydroxyethyl)-3-methylimidazolium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Certificates
About this item
CAS Number:174899-86-6
:
Formula:C₈H₁₁F₆N₃O₅S₂
MDL Number:MFCD09038874
:407.30 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %