Order Entry
Puerto Rico
ContactUsLinkComponent
5,5'''-Didodecyl-2,2':5',2'':5'',2'''-quaterthiophene ≥96.0% (by HPLC)
5,5'''-Didodecyl-2,2':5',2'':5'',2'''-quaterthiophene ≥96.0% (by HPLC)
Catalog # TCD4889-100MG
Supplier:  TCI America
CAS Number:  
5,5'''-Didodecyl-2,2':5',2'':5'',2'''-quaterthiophene ≥96.0% (by HPLC)
Catalog # TCD4889-100MG
Supplier:  TCI America
Supplier Number:  D4889-100MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 mg
  • Cat. No.:
    TCD4889-100MG

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Elemental analysis (Sulfuride):
    18.20 to 20.20 %

Specification Test Results

About this item

 :
Melting Pt:166°C
 :667.14 g/mol
CAS Number:153561-79-6
Formula:C₄₀H₅₈S₄