Order Entry
Puerto Rico
ContactUsLinkComponent
[4-(Bromomethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate ≥90.0% (by HPLC)
Irritant
[4-(Bromomethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate ≥90.0% (by HPLC)
Catalog # 76219-550
Supplier:  TCI America
CAS Number:  
Irritant
[4-(Bromomethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate ≥90.0% (by HPLC)
Catalog # 76219-550
Supplier:  TCI America
Supplier Number:  B5269-200MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    200 mg
  • Cat. No.:
    76219-550

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 90.0 % (total of Ion-Pair)

Specification Test Results

About this item

 :
 :635.31 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1453864-75-9
Formula:C₂₃H₂₄BrIO₆S