Order Entry
Puerto Rico
ContactUsLinkComponent
Brompheniramine maleate ≥98.0% (by HPLC, titration analysis)
Irritant
Brompheniramine maleate ≥98.0% (by HPLC, titration analysis)
Catalog # TCB5167-25G
Supplier:  TCI America
CAS Number:  
Irritant
Brompheniramine maleate ≥98.0% (by HPLC, titration analysis)
Catalog # TCB5167-25G
Supplier:  TCI America
Supplier Number:  B5167-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    25 g
  • Cat. No.:
    TCB5167-25G

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 % (total of Ion-Pair)
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    134.0 to 138.0 deg-C

Specification Test Results

About this item

 :
 :319.24 g/mol
Merck Index:14,01443
EINECS:213-562-9
 :GHS07
CAS Number:980-71-2
Formula:C₁₆H₁₉BrN₂
MDL Number:MFCD00865691