Order Entry
Puerto Rico
ContactUsLinkComponent
(5-Carboxypentyl)triphenylphosphonium bromide ≥96.0% (by titrimetric analysis)
Irritant
(5-Carboxypentyl)triphenylphosphonium bromide ≥96.0% (by titrimetric analysis)
Catalog # 76218-904
Supplier:  TCI America
CAS Number:  
Irritant
(5-Carboxypentyl)triphenylphosphonium bromide ≥96.0% (by titrimetric analysis)
Catalog # 76218-904
Supplier:  TCI America
Supplier Number:  C3113-5G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    5 g
  • Cat. No.:
    76218-904

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Neutralization titration):
    min. 96.0 %
  • Melting point:
    198.0 to 202.0 deg-C

Specification Test Results

About this item

 :
 :457.36 g/mol
 :GHS07
CAS Number:50889-29-7
Formula:C₂₄H₂₆BrO₂P
MDL Number:MFCD00055556