

1-Butyl-4-methylpyridiniumtetrafluoroborate ≥98.0% (by HPLC)
Supplier: TCI America
Certificates
About this item
CAS Number:343952-33-0
:
Formula:C₁₀H₁₆BF₄N
MDL Number:MFCD03095460
:237.05
Melting Pt:-30°C
UN:1760
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %