
1-Benzyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Certificates
About this item
CAS Number:36443-80-8
:
Formula:C₁₁H₁₃ClN₂
MDL Number:MFCD07784451
:208.69
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together