Order Entry
Puerto Rico
ContactUsLinkComponent
Tellurium dioxide ≥98.0% (W)
Irritant
Health
Tellurium dioxide ≥98.0% (W)
Catalog # TCT3254-25G
Supplier:  TCI America
CAS Number:  
Irritant
Health
Tellurium dioxide ≥98.0% (W)
Catalog # TCT3254-25G
Supplier:  TCI America
Supplier Number:  T3254-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    25 g
  • Cat. No.:
    TCT3254-25G

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Gravimetric Method):
    min. 98 %

Specification Test Results

About this item

 :
 :159.6 g/mol
Merck Index:14,09123
Melting Pt:733°C
EINECS:231-193-1
 :GHS07
 :GHS08
CAS Number:7446-07-3
Formula:O₂Te
MDL Number:MFCD00011263