


Boron trichloride ∼13% ∼1.0 mol/L in toluene
Catalog # TCB4415-100ML
Supplier: TCI America
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:100 ml
- Cat. No.:TCB4415-100ML
- Supplier no.:B4415-100ML
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Concentration (Neutralization titration):12.5% to 14.0 w/w%
Specification Test Results
About this item
:
:117.17
Merck Index:14,01348
Storage Temperature:Refrigerator
EINECS:233-658-4
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS08
CAS Number:10294-34-5
Formula:BCl₃
MDL Number:MFCD00011313