Order Entry
Puerto Rico
ContactUsLinkComponent
Boron trichloride ∼13% ∼1.0 mol/L in p-xylene
Flammable
Corrosive
Irritant
Health
Boron trichloride ∼13% ∼1.0 mol/L in p-xylene
Catalog # TCB3968-100ML
Supplier:  TCI America
CAS Number:  
Flammable
Corrosive
Irritant
Health
Boron trichloride ∼13% ∼1.0 mol/L in p-xylene
Catalog # TCB3968-100ML
Supplier:  TCI America
Supplier Number:  B3968-100ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 ml
  • Cat. No.:
    TCB3968-100ML
  • Supplier no.:
    B3968-100ML

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Concentration (Neutralization titration):
    12.5 to 14.5 w/w%

Specification Test Results

About this item

 :
 :117.17
Merck Index:14,01348
Flash Pt:29°C
Storage Temperature:Refrigerator
EINECS:233-658-4
UN:2920
ADR:8,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:10294-34-5
Formula:BCl₃
MDL Number:MFCD00011313