LabExact® Nylon Membranes, Non sterile, Hydrophilic
Supplier: I.W. TREMONT CO., INC.
New Product
Certificates
About this item
LabExact® Nylon Membrane is supported with a polyethylene terephthalate (PET) scrim for added support without using a bonding agent to ensure purity, chemical compatibility, and lower extractables.
- Hydrophilic material eliminates need for wetting agents
- Strong protein bonding with high recoveries
- Autoclavable
This scrim is bonded to the nylon using a unique technique with no bonding agent for purity, chemical compatibility, and lower extractables.
Packaging: Box of 100.
Frequently Bought Together