ContactUsLinkComponent
LabExact® CFP542 Quantitative Ashless Hardened Cellulose Filter Papers, 2 to 3 µm
LabExact® CFP542 Quantitative Ashless Hardened Cellulose Filter Papers, 2 to 3 µm
New Product
Certificates
LabExact® CFP542 Quantitative Ashless Hardened Cellulose Filter Papers, 2 to 3 µm
New Product
Specifications
  • Catalog No:
  • 77590-758
  • 77590-756
  • 77590-754
  • 77590-752
  • 77590-750
  • 77590-748
  • Ash Content:
  • <0.01%
  • <0.01%
  • <0.01%
  • <0.01%
  • <0.01%
  • <0.01%
  • Basis Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Retention:
  • 2 - 3 µm
  • 2 - 3 µm
  • 2 - 3 µm
  • 2 - 3 µm
  • 2 - 3 µm
  • 2 - 3 µm