Fire Blanket, Sellstrom®
33000-159
Small Business Enterprise
- Description:Fire Blanket
- Cat. No.:33000-159
- Supplier no.:97450
Washable, lightweight, woven fiberglass emergency fire blanket is designed to prevent further fire accidents in the lab.
- Sparks, flash fires, and flying particles can be easily extinguished
- Adequately sized to wrap a fire victim
- To clean, wash with soft sponge and air dry only
- Sewn-on grommets allow for hanging during cleaning process
Complete with red herculite pouch for wall hanging. Velcro® pull tab on bottom releases 1.5×1.8 m (5×6') blanket.