Portable Eyewash, Speakman®
Portable Eyewash
Catalog # VF-0590-SE
Supplier: Speakman
Supplier #: SE-590
Jump to:
Product Details & Documents
Perfect for environmental field labs, this portable eyewash and drench hose can be used where permanent water supply is not available.
- Complete with pressure gauge, air filter, and pressure relief valve
- Stainless steel tank has an 30.3 L (8 gal.) water capacity
Features twin chrome-plated eyewash spray heads activated by a stainless steel push handle. The stay-open valve releases 1.5 L/min. (0.4 gpm) spray for 15 minutes. The 1.8 m (6') long reinforced drench hose features a self-closing squeeze valve.
Specifications
- Description:Portable Eyewash