GRAVITYFLO® Gravity-Fed Eye/Face Wash, Speakman®
VF-4000-SE
: Speakman
Small Business Enterprise
- Description:GRAVITYFLO* Gravity-Fed Eye/Face Wash
- Cat. No.:VF-4000-SE
- Supplier no.:SE-4000
This portable, gravity-fed eye/face wash is perfect for environmental site locations as well as areas that are not plumbed.
- Easy-carry molded hand grips are located in the water head
- Can be wall mounted with a bracket (included) or shelf mounted
Constructed of durable high density plastic, this 60.6 L (16 gal.) unit features large rubber spray heads for complete eye and face coverage. Open the quick-acting valve by pulling the spray arms down. In the 'off' position, spray heads are enclosed in a recessed area for protection from contamination.
: Meets ANSI Z358-1 standard for portable eye wash units. Meets ANSI A117.1 standard for access and use by physically handicapped people.