Calibrated Micropipettes, Drummond Scientific
Supplier: Drummond
Certificates
About this item
Borosilicate glass micropipettes are ideal for a variety of liquid transfer work.
- Color-coded micropipettes with imprinted sizes
- Fire polished
- Accuracy of ±0.5%
- Anticoagulant treatment for clinical applications (special limited sizes)
The fire-polished micropipettes offer ±0.5% accuracy.
Ideal for a variety of liquid transfer work, 250 color-coded micropipettes and one 15" aspirator tube are supplied in an easy-to-store cylinder.
Ordering information: Packs include 250 micropipettes with one 38.1 cm (15") rubber aspirator tube supplied in a conveniently designed cylinder. Sizes of Precise Calibrated Micropipettes range from one to 200 µl. For blood transfer work, a heparinized 44.7 µl pipette is also available.
Frequently Bought Together