VWR® Bead Baths
About this item
The VWR® Bead bath series is ideal for precision incubation of laboratory samples. Unlike traditional water baths, these baths can also accurately maintain temperature of thermal beads (such as LabArmor® brand) proving a wide range of benefits.
- Eliminates constant refills
- Accessory free tube/vessel loading
- Reduces the risk of splash contamination
- Facilitate angled tube incubations
- Precision temperature control
- Accepts beads or water
- In-lab calibration feature
Two sizes are available, including a standard size 10 L model and a mini sized 2 L model. Each model includes a transparent cover for reduced energy consumption, while providing constant visualization of samples.
For precision temperature control, can be used with thermal beads (such as LabArmor® brand) or water.
Temperature accuracy: ±0.25 °C
Certifications: CE marked.
Ordering information: Thermal beads must be ordered separately.
Delivery information: Supplied with a transparent cover.
2 Options Available Below
Product Details & Documents
The VWR® Bead bath series is ideal for precision incubation of laboratory samples. Unlike traditional water baths, these baths can also accurately maintain temperature of thermal beads (such as LabArmor® brand) proving a wide range of benefits.
- Eliminates constant refills
- Accessory free tube/vessel loading
- Reduces the risk of splash contamination
- Facilitate angled tube incubations
- Precision temperature control
- Accepts beads or water
- In-lab calibration feature
Two sizes are available, including a standard size 10 L model and a mini sized 2 L model. Each model includes a transparent cover for reduced energy consumption, while providing constant visualization of samples.
For precision temperature control, can be used with thermal beads (such as LabArmor® brand) or water.
Temperature accuracy: ±0.25 °C
Certifications: CE marked.
Ordering information: Thermal beads must be ordered separately.
Delivery information: Supplied with a transparent cover.
Product Family Options
Product Information
- CapacityElectricalTemperature rangeUniformityDimensionsAvailabilityPrice
- 77553-322VWR® bead bath, chamber size: 13.7×15.2×10.2 cm (5.4×6×4"), with 1 °C temperature increments, weight 2.2 kg (4.8 lbs.)VWR® bead bath, chamber size: 13.7×15.2×10.2 cm (5.4×6×4"), with 1 °C temperature increments, weight 2.2 kg (4.8 lbs.)2 L115 VAmbient +5...70 °C (Bead)
Ambient +5...80 °C (Water)±1.0 °C16.8×20×15 cm (6.6×8×5.9")Log in for availabilityEach$1,013.52 - 77553-324VWR® bead bath, chamber size: 24.1×29.7×15 cm (9.5×11.7×5.9"), with 0.1 °C temperature increments, weight 6.8 kg (15 lbs.)VWR® bead bath, chamber size: 24.1×29.7×15 cm (9.5×11.7×5.9"), with 0.1 °C temperature increments, weight 6.8 kg (15 lbs.)10 L115 VAmbient +5...70 °C (Bead)
Ambient +5...80 °C (Water)±1.0 °C27.4×41.1×30.5 cm (10.8×16.2×12")Log in for availabilityEach$1,728.34
Beads
Product Information
- Catalog #SizeØAvailabilityPrice