Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein B100
- Antigen Symbol:APOB100
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:338
- Antigen Synonyms:Apo-B100
- Amino Acid Number:3365 - 3548
- UniProtKB:P04114
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:26 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant APOB100(His3365~Glu3548)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA603HU1
- Packaging:Vial
Specifications
About this item
Polyclonal antibody to Apolipoprotein B100 (APOB100), derived from recombinant APOB100(His3365~Glu3548), is reactive with Human.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
Apolipoprotein B100 is a protein that plays a role in metabolism. It is a form of low density lipoprotein (LDL). Apo-B levels correlate with the risk of coronary disease. The protein occurs in the plasma in 2 main isoforms, APOB48 and APOB100. The first is synthesized exclusively by the small intestine, the second by the liver. Both isoforms are coded by APOB and by a single mRNA transcript larger than 16 kb. Apo-B100 is the major physiological ligand for the LDL receptor. Apo-B100 is a large monomeric protein, containing 4536 amino acids. Apo-B100 is synthesized in the liver and is required for the assembly of VLDL. It does not interchange between lipoprotein particles, as do the other apolipoproteins, and it is found in IDL and LDL after the removal of the Apo-A, E and C. Apo-B48 is present in chylomicrons and their remnants. It is essential for the intestinal absorption of dietary lipids.
Type: Primary
Antigen: APOB100
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human