Specifications
- Antibody Type:Primary
- Antigen Name:Relaxin/Insulin Like Family Peptide Receptor 1
- Antigen Symbol:RXFP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:59350
- Antigen Synonyms:RXFP1|Leucine-Rich Repeat-Containing G Protein-Coupled Receptor 7|LGR7.1|LGR7|Relaxin family peptide receptor 1|RXFPR1|LGR7.2|Relaxin receptor 1|LGR7.10
- Amino Acid Number:260 - 409
- UniProtKB:Q9HBX9
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:17 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant RXFP1(Pro260~Met409)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB456HU1
- Packaging:Vial
Specifications
About this item
Polyclonal antibody to Relaxin/Insulin Like Family Peptide Receptor 1 (RXFP1), derived from recombinant RXFP1(Pro260~Met409), is reactive with Human/Mouse.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
The deduced 757- and 723-amino acid LGR7 proteins are 33% identical to snail Lgr. They contain a low density lipoprotein receptor cysteine-rich motif, 10 LRRs, and 5 N-glycosylation sites in the N terminus, the 7-transmembrane region, and a unique C-terminal tail with a phosphorylation site. Northern blot analysis revealed expression of a 5.5-kb transcript in all rat tissues tested except spleen, and smaller transcripts were found in oviduct, uterus, colon, and brain. Expression of the long and short variants of LGR7 with mutations in transmembrane domain 6 (asp637 to tyr and asp603 to tyr, respectively), but not of wildtype LGR7, resulted in increased basal production of cAMP, as seen in similar experiments with the luteinizing hormone receptor (LHCGR) and the TSH receptor but not the FSH receptor.
Type: Primary
Antigen: RXFP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse