Specifications
- Antibody Type:Primary
- Antigen Name:FMS Like Tyrosine Kinase 3 Ligand
- Antigen Symbol:Flt3L
- Clonality:Monoclonal
- Clone:D4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:2323
- Antigen Synonyms:FLT3-LG|SL cytokine|FLT3LG|Fms-Related Tyrosine Kinase 3 ligand|FL|Flt3-L
- Amino Acid Number:83 - 182
- UniProtKB:P49771
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:43 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant Flt3L(Met83~Pro182)
- Purification:Protein A + Protein G affinity chromatography
- Cat. No.:MSPP-MAA038HU2
- Packaging:Vial
Specifications
About this item
Monoclonal antibody to FMS Like Tyrosine Kinase 3 Ligand (Flt3L), derived from recombinant Flt3L(Met83~Pro182), is reactive with Human.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
The Flt3l is a small molecule that acts as a growth factor that increases the number of immune cells by activating the hematopoietic progenitors. It also induces the mobilization of the hematopoietic progenitors and stem cells in vivo which may help the system to kill cancer cells. FLT3L is crucial for steady-state pDC and cDC development. A lack of FLT3L results in low levels of DCs. Flt3 ligand (FL) is a hematopoietic four helical bundle cytokine. It is structurally homologous to stem cell factor (SCF) and colony stimulating facor 1 (CSF-1). In synergy with other growth factors, Flt3 ligand stimulates the proliferation and differentiation of various blood cell progenitors.
Type: Primary
Antigen: Flt3L
Clonality: Monoclonal
Clone: D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human