Specifications
- Antibody Type:Primary
- Antigen Name:Cathepsin Z
- Antigen Symbol:CTSZ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:1522
- Antigen Synonyms:CTS-Z|Cathepsin X|Cathepsin P|CTSX
- Amino Acid Number:24 - 303
- UniProtKB:Q9UBR2
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:38 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant CTSZ(Gly24~Val303)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAC969HU1
- Packaging:Vial
Specifications
About this item
Polyclonal antibody to Cathepsin Z (CTSZ), derived from recombinant CTSZ(Gly24~Val303), is reactive with Human/Rat.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
Cathepsin Z or cathepsin X is a lysosomal cysteine proteinase and member of the peptidase C1 family. It exhibits both carboxy-monopeptidase and carboxy-dipeptidase activities. The encoded protein has also been known as cathepsin X and cathepsin P. This gene is expressed ubiquitously in cancer cell lines and primary tumors and, like other members of this family, may be involved in tumorigenesis. The predicted 303-amino acid protein has the domain structure characteristic of cysteine proteinases, including a signal sequence, a prodomain, and a catalytic domain responsible for proteolytic activity. However, it also contains 3 unique peptide insertions. Cathepsin Z shares protein sequence identity with other human cysteine proteinases of the papain family, including 34% with cathepsin C and 26% with cathepsin B.
Type: Primary
Antigen: CTSZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat