Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-VB6 Rabbit Polyclonal Antibody
  MSPP-PAA916GE1
Anti-VB6 Rabbit Polyclonal Antibody
  MSPP-PAA916GE1
 :  PAA916GE01

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Vitamin B6
  • Antigen Symbol:
    VB6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Multiple
  • Size:
    200 µl
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Antigen Synonyms:
    Pyridoxine
  • Storage Buffer:
    0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
  • Molecular Weight:
    169.2Da
  • Storage Temperature:
    Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
  • Concentration:
    1 mg/ml (for reference only)
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    OVA conjugated VB6
  • Purification:
    Antigen-specific affinity chromatography followed by Protein A affinity chromatography
  • Cat. No.:
    MSPP-PAA916GE1
  • Packaging:
    Vial

 

 

Polyclonal antibody to Vitamin B6 (VB6), derived from OVA conjugated VB6, is reactive with General species.

  • High specificity, affinity and purity
  • Extensive validation and batch-to-batch consistency
  • Diverse range of conjugated and custom options
  • Comprehensive validation data and documentation
  • All in stock and fast global delivery
  • 100% Quality and service satisfaction guarantee!

Vitamin B6 is a water-soluble vitamin. Pyridoxine is one of the compounds that can be called vitamin B6, along with pyridoxal and pyridoxamine. Its hydrochloride salt pyridoxine hydrochloride is often used.  Vitamin B6 assists in the balancing of sodium and potassium as well as promoting red blood cell production. It is linked to cardiovascular health by decreasing the formation of homocysteine. Pyridoxine may help balance hormonal changes in women and aid the immune system. It is required for the production of the monoamine neurotransmitters serotonin, dopamine, norepinephrine and epinephrine, as it is the precursor to pyridoxal phosphate: cofactor for the enzyme aromatic amino acid decarboxylase. This enzyme is responsible for converting the precursors 5-hydroxytryptophan (5-HTP) into serotonin and melatonin, and levodopa (L-DOPA) into dopamine, noradrenaline and adrenaline.

 : For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Type: Primary
Antigen: VB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species