Specifications
- Antibody Type:Primary
- Antigen Name:Phosphoglycerate Kinase 1
- Antigen Symbol:PGK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:5230
- Antigen Synonyms:PGKA|Cell migration-inducing gene 10 protein|MIG10|Primer recognition protein 2|PRP 2
- Amino Acid Number:2 - 417
- UniProtKB:P00558
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:48 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant PGK1(Ser2~Ile417)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAG304HU1
- Packaging:Vial
Specifications
About this item
Polyclonal antibody to Phosphoglycerate Kinase 1 (PGK1), derived from recombinant PGK1(Ser2~Ile417), is reactive with Human/Mouse.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
The Phosphoglycerate Kinase 1 is a glycolytic enzyme that catalyzes the conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate. The encoded protein may also act as a cofactor for polymerase alpha. This gene lies on the X-chromosome, while a related pseudogene also has been found on the X-chromosome and another on chromosome 19.PGK1 is distinguished from testicular PGK2, which maps to chromosome 6p21.
The deduced protein contains 417 amino acid residues. Southern blot analysis of human genomic DNAs showed a complex pattern of hybridizing fragments, 2 of which were non-X in origin. The results were interpreted as reflecting the existence of a small family of dispersed PGK or PGK-like genes.
Type: Primary
Antigen: PGK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse