Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor, Ionotropic, AMPA 3
- Antigen Symbol:GRIA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:2892
- Antigen Synonyms:gluR-C|GLUR3|AMPA-selective glutamate receptor 3|GLURC|GLUR-K3
- Amino Acid Number:649 - 823
- UniProtKB:P42263
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:23 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant GRIA3(Thr649~Asn823)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAE803HU1
- Packaging:Vial
Specifications
About this item
Polyclonal antibody to Glutamate Receptor, Ionotropic, AMPA 3 (GRIA3), derived from recombinant GRIA3(Thr649~Asn823), is reactive with Human/Mouse/Rat/Pig.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes with multiple subunits, each possessing transmembrane regions, and all arranged to form a ligand-gated ion channel. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. Glutamate receptor 3 belongs to a family of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate (AMPA) receptors. Alternative splicing at this locus results in several different isoforms which may vary in their signal transduction properties.
Type: Primary
Antigen: GRIA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig