To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Octopamine
- Antigen Symbol:OA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Size:200 µl
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:β,4-Dihydroxyphenethylamine|Norfen|Norden|Epirenor
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:153.2g/mol
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:OVA conjugated OA
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAG048GE1
- Packaging:Vial
Specifications
About this item
Polyclonal antibody to Octopamine (OA), derived from OVA conjugated OA, is reactive with General species.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
Octopamine also known as β,4-dihydroxyphenethylamine, is an endogenous biogenic amine that is closely related to norepinephrine, and has effects on the adrenergic and dopaminergic systems. Biosynthesis of the D(-)-enantiomer of octopamine is by β-hydroxylation of tyramine via the enzyme dopamine β-hydroxylase. Under the trade names Epirenor, Norden, and Norfen, octopamine is also used clinically as a sympathomimetic agent. Octopamine was first discovered by Italian scientist Vittorio Erspamer in 1948 in the salivary glands of the octopus and has since been found to act as neurotransmitter, neurohormone and neuromodulator in invertebrates. It is widely used in energy-demanding behaviours by all insects, crustaceans (crabs, lobsters, crayfish) and spiders. Such behaviours include flying, egg-laying, and jumping.
Type: Primary
Antigen: OA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species