Specifications
- Antibody Type:Primary
- Antigen Name:Keratin 25
- Antigen Symbol:KRT25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:70810
- Antigen Synonyms:K25|Keratin|Cytokeratin-25|Type I inner root sheath-specific keratin-K25irs1|KRT25A|type I cytoskeletal 25|CK-25
- Amino Acid Number:1 - 446
- UniProtKB:Q8VCW2
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:53 kDa
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant KRT25(Met1~Asn446)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAS211MU1
- Packaging:Vial
Specifications
About this item
Polyclonal antibody to Keratin 25 (KRT25), derived from recombinant KRT25(Met1~Asn446), is reactive with Mouse.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
KRT25 encodes a member of the type I (acidic) keratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. Strongly expressed in skin and scalp. In the hair follicle, expressed in Henle layer, Huxley layer and in the inner root sheath cuticle of the hair follicle. Expression extends from the bulb region up to the point of differentiation into the three layers. Also present in the medulla of beard hair (at protein level). Essential for the proper assembly of type I and type II keratin protein complexes and formation of keratin intermediate filaments in the inner root sheath (irs)
Type: Primary
Antigen: KRT25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse