- Antibody Type:Primary
- Antigen Name:Procollagen I N-Terminal Propeptide
- Antigen Symbol:PINP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:12842
- Antigen Synonyms:N-Propeptide Of Type I Procollagen|P1NP|Procollagen I Amino Terminal Propeptide
- UniProtKB:P11087
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Storage Temperature:Store at 2...8 °C for frequent use, −20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:OVA conjugated PINP
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA957MU1
- Packaging:Vial
Polyclonal antibody to Procollagen I N-Terminal Propeptide (PINP), derived from OVA conjugated PINP, is reactive with Mouse.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee!
The PINP molecule is similar to PIIINP consisting of three distinct structural domains: Col 1 is on the aminoterminal side of the molecule, while Col 2 and Col 3 are situated on the middle of the helically structured molecule (Kühn et al. 1982). The PINP molecule has a molecular mass of 35000 and is cleared by scavenger receptors in liver endothelial cells (Melkko et al. 1994). PINP often occurs in circulation in two forms of different molecular sizes. One is identical to the trimeric authentic antigen (intact PINP) whereas the other consists of smaller forms of PINP, resembling a single domain of the proα1(I) chain of PINP and is probably a degradation product of type I procollagen or I pN-collagen. Thus, an assay of intact PINP rather than total PINP appears to be more sensitive in detecting changes in the rate of type I collagen synthesis (Melkko et al. 1996, Risteli and Risteli 1999).
Type: Primary
Antigen: PINP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse