Anti-PACAP Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Polyclonal Antibody to Pituitary Adenylate Cyclase Activating Peptide (PACAP), derived from recombinant PACAP (Ile17~Leu176), is reactive with Human/Mouse.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Pituitary adenylate cyclase-activating peptide, adenylate cyclase-activating polypeptide 1. Mediated by adenylate cyclase-activating polypeptide 1 receptors, this polypeptide stimulates adenylate cyclase and subsequently increases the cAMP level in target cells. Adenylate cyclase-activating polypeptide 1 not only is a hypophysiotropic hormone but also functions as a neurotransmitter and neuromodulator. In addition, it plays a role in paracrine and autocrine regulation of certain types of cells. This gene is composed of five exons. Exons 1 and 2 encode the 5' UTR and signal peptide, respectively; exon 4 encodes an adenylate cyclase-activating polypeptide 1-related peptide; and exon 5 encodes the mature peptide and 3' UTR.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.
Type: Primary
Antigen: PACAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Documents
Cloud-Clone Quality Guarantee
Additional Product Details
Specifications
- Size:200 µl
- Packaging:Vial
- Cross Adsorption:No
- Clonality:Polyclonal
- Molecular Weight:47 kDa
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Form:Liquid
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- UniProtKB:P18509
- Isotype:IgG
- Host:Rabbit
- Concentration:0.5 mg/ml (for reference only)
- Cat. No.:MSPP-PAB347HU1
- Reactivity:Human|Mouse
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice at 4 °C
- Antibody Type:Primary
- Antigen Symbol:PACAP
- Gene ID:116
- Immunogen:recombinant PACAP (Ile17~Leu176)
- ImmunoChemistry:Yes
- Amino Acid Number:17 - 176
- Conjugation:Unconjugated
- Antigen Synonyms:PRP-48|ADCYAP1|Pituitary Adenylate Cyclase Activating Peptide
- Antigen Name:Pituitary Adenylate Cyclase Activating Peptide
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.



