Specifications
- Antibody Type:Primary
- Antigen Name:Breast Cancer Susceptibility Protein 2
- Antigen Symbol:BRCA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:12190
- Antigen Synonyms:FAD|FANCD|BRCA2|FANCD1|FACD|Breast Cancer 2,Early Onset|FAD1|Breast Cancer Type 2 Susceptibility Protein|Fanconi anemia group D1 protein|BRCC2
- Amino Acid Number:2990 - 3232
- UniProtKB:P97929
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:30 kDa
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:recombinant BRCA2 (Glu2990~Ala3232)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB364MU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Breast Cancer Susceptibility Protein 2 (BRCA2), derived from recombinant BRCA2 (Glu2990~Ala3232), is reactive with Mouse.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
BRCA2 is a protein belongs to the tumor suppressor gene family and the protein encoded by this gene is involved in the repair of chromosomal damage with an important role in the error-free repair of DNA double strand breaks. The BRCA2 gene is located on the long (q) arm of chromosome 13 at position 12.3 (13q12.3), from base pair 31, 787, 616 to base pair 31, 871, 804. The BRCA2 protein binds to and regulates the protein produced by the RAD51 gene to fix breaks in DNA. These breaks can be caused by natural and medical radiation or other environmental exposures, but also occur when chromosomes exchange genetic material during a special type of cell division that creates sperm and eggs (meiosis).
Type: Primary
Antigen: BRCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse