Specifications
- Antibody Type:Primary
- Antigen Name:Adenosine Triphosphate
- Antigen Symbol:ATP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:Adenosine-5'-Triphosphate
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:507.2 Da
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.76 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:OVA conjugated ATP
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA349GE1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Adenosine Triphosphate (ATP), derived from OVA conjugated ATP, is reactive with General species.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Adenosine-5'-triphosphate (ATP) is a multifunctional nucleotide that plays an important role in cell biology as a coenzyme, that is, the 'molecular unit of currency' of intracellular energy transfer. ATP transports chemical energy within cells for metabolism. It is an energy source produced during photosynthesis and cellular respiration and consumed by many enzymes and a multitude of cellular processes, including biosynthetic reactions, motility, and cell division. ATP is made from adenosine diphosphate (ADP) or adenosine monophosphate (AMP) and its use in metabolism converts it back into these precursors. ATP is therefore continuously recycled in organisms, with the human body turning over its own weight in ATP each day. ATP was discovered in 1929 by Karl Lohmann, but its correct structure was not determined until some years later. It was proposed to be the main energy-transfer molecule in the cell by Fritz Albert Lipmann in 1941.
Type: Primary
Antigen: ATP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species