Specifications
- Antibody Type:Primary
- Antigen Name:Apelin
- Antigen Symbol:APLN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:30878
- Antigen Synonyms:XNPEP2|APJ endogenous ligand|APEL
- Amino Acid Number:23 - 77
- UniProtKB:Q9R0R4
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:39 kDa
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.3 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:recombinant APLN (Val23~Phe77)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAD066MU1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Apelin (APLN), derived from recombinant APLN (Val23~Phe77), is reactive with Mouse.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Apelin also known as APLN is a peptide which is encoded by the APLN gene. Apelin is the endogenous ligand for the G-protein-coupled APJ receptor that is expressed at the surface of some cell types. It is widely expressed in various organs such as the heart, lung, kidney, liver, adipose tissue, gastrointestinal tract, brain, adrenal glands, endothelium, and human plasma. The gene encodes a 77-amino acid polypeptide that includes a secretory signal sequence. Synthetic 36-amino acid apelin (apelin-36), corresponding to the most C-terminal 36 residues of preproapelin, caused the acidification of cells expressing transfected APJ, but not other receptors. Apelin is an endogenous ligand for the APJ receptor.
Type: Primary
Antigen: APLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse