Specifications
- Antibody Type:Primary
- Antigen Name:Natural Cytotoxicity Triggering Receptor 1
- Antigen Symbol:NCR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:17086
- Antigen Synonyms:CD335|NCR1|NK cell-activating receptor|Lymphocyte antigen 94 homolog|Natural killer cell p46-related protein|LY94|NK-P46|NKP46
- Amino Acid Number:17 - 211
- UniProtKB:Q8C567
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:26 kDa
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:recombinant NCR1 (Gln17~Thr211)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB80MU01
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Natural Cytotoxicity Triggering Receptor 1 (NCR1), derived from recombinant NCR1 (Gln17~Thr211), is reactive with Mouse/Rat.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Natural cytotoxicity triggering receptor 1 is a protein encoded by the NCR1 gene. By screening a human NK cell cDNA library, Pessino et al., (1998) cloned the cDNA for NKp46, or LY94, an Ig superfamily member that may cooperate with other activating receptors. The LY94 cDNA encodes a 304-amino acid type I transmembrane protein. The extracellular region is preceded by a 21-residue signal peptide with 2 cysteine-bridged C2-type Ig-like domains. A stem connects the extracellular domain to the apparently short transmembrane domain of 19 amino acids, which includes an arginine. The intracellular region contains 30 amino acids and is rich in basic residues, but does not contain any immunoreceptor tyrosine-based activating motifs (ITAM).
Type: Primary
Antigen: NCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat