Specifications
- Antibody Type:Primary
- Antigen Name:Alpha-1-B-Glycoprotein
- Antigen Symbol:a1BG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:117586
- Amino Acid Number:22 - 219
- UniProtKB:Q19LI2
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:31 kDa
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.5 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:recombinant a1BG (Met22~Ser219)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAE570MU3
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Alpha-1-B-Glycoprotein (a1BG), derived from recombinant a1BG (Met22~Ser219), is reactive with Mouse/Human/Rat.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Alpha-1B-glycoprotein is present in normal adult plasma at an average concentration of 22 mg/dl. Gahne et al., (1987) observed genetic polymorphism of A1B using one-dimensional horizontal polyacrylamide gel electrophoresis followed by Western blotting with specific antiserum. Three different phenotypes, designated 1-1, 1-2, and 2-2, were observed. Family data supported the hypothesis that the three phenotypes are determined by 2 codominant alleles at an autosomal locus. In pigs the homologous locus is linked to malignant hyperthermia. Several other linkages in pigs and in horses suggest that human chromosomes 19, 6, and 1 are 'candidate chromosomes' for bearing the human A1B.
Type: Primary
Antigen: a1BG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human, Rat