- Antibody Type:Primary
- Antigen Name:Parathyroid Hormone
- Antigen Symbol:PTH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Gene ID:24694
- Antigen Synonyms:Parathyrin|Parathormone|Intact Parathyroid Hormone|iPTH
- Amino Acid Number:32 - 115
- UniProtKB:P04089
- Storage Buffer:PBS, pH 7.4, containing 0.02% NaN₃, 50% glycerol
- Molecular Weight:59 kDa
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:recombinant PTH (Ala32~Gln115)
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA866RA1
- Packaging:Vial
Polyclonal Antibody to Parathyroid Hormone (PTH), derived from recombinant PTH (Ala32~Gln115), is reactive with Rat.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Parathyroid hormone is secreted by the parathyroid glands as a polypeptide containing 84 amino acids. It acts to increase the concentration of calcium (Ca²⁺) in the blood, whereas calcitonin (a hormone produced by the parafollicular cells (C cells) of the thyroid gland) acts to decrease calcium concentration. PTH acts to increase the concentration of calcium in the blood by acting upon parathyroid hormone receptor in three parts of the body: PTH half-life is approximately 4 minutes. It has a molecular mass of 9.4 kDa. PTH was one of the first hormones to be shown to use the G-protein, adenylyl cyclase second messenger system. Normal total plasma calcium level ranges from 8.5 to 10.2 mg/dl (2.12 mmol/L to 2.55 mmol/L).
Type: Primary
Antigen: PTH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat