To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Asymmetrical Dimethylarginine
- Antigen Symbol:ADMA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:NG,NG-Dimethylarginine dihydrochloride|N,N-Dimethylarginine|Asymmetric Dimethylarginine
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:202.3 Da
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.45 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:BSA conjugated ADMA
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAB301GE1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to Asymmetrical Dimethylarginine (ADMA), derived from BSA conjugated ADMA, is reactive with General species.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Asymmetric dimethylarginine (ADMA) is a naturally occurring chemical found in blood plasma. It is a metabolic by-product of continual protein modification processes in the cytoplasm of all human cells. It is closely related to L-arginine, a conditionally-essential amino acid. ADMA interferes with L-arginine in the production of nitric oxide, a key chemical involved in normal endothelial function and, by extension, cardiovascular health. Nitric oxide (NO) is synthesized in endothelial cells by the enzyme endothelial nitric oxide synthase. The production rate of NO is a key factor for endothelial and cardiovascular functions. The substrate of endothelial nitric oxide synthase is Arg, which is converted into the products NO and citrulline. The enzyme is inhibited by the endogenous Arg metabolite asymmetric dimethylarginine (ADMA).
Type: Primary
Antigen: ADMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species